4-Fluoroanthranilic acid ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI
Varning

Synonyms: 2-Amino-4-fluorobenzoic acid

F0405-1G F0405-25G F0405-5G
TCIAF0405-1GEA 495 SEK
TCIAF0405-1G TCIAF0405-25G TCIAF0405-5G
4-Fluoroanthranilic acid ≥98.0% (by HPLC, titration analysis)
4-Fluoroanthranilic acid
Formel: C₇H₆FNO₂
Kokpunkt: 305 °C (1013 hPa)
Smältpunkt: 198…199 °C
Storage Temperature: Omgivande
MDL Number: MFCD00075553
CAS nummer: 446-32-2
EINECS: 207-163-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20716acc8f8b346ab34bdabff9c0a09fbbc97434&vid=dcad2c45d1ed4a3803a39d98183d6a5e844f41cf&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR