Pentafluorobenzoic acid ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 2,3,4,5,6-Pentafluorobenzoic acid

P0806-25G P0806-5G
TCIAP0806-25GEA 765 SEK
TCIAP0806-25G TCIAP0806-5G
Pentafluorobenzoic acid ≥98.0% (by GC, titration analysis)
Pentafluorobenzoic acid
Formel: C₇HF₅O₂
MW: 212,08 g/mol
Kokpunkt: 220 °C (1013 hPa)
Smältpunkt: 100…102 °C
Densitet: 1,942 g/cm³ (22 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002406
CAS nummer: 602-94-8
EINECS: 210-026-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27f15adafe7dea175d110fa4db1060bb35016ca6&vid=4eb4908389940af672589c44018e379c801e2773&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 103.0 to 105.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR