3-Fluorobenzoic acid ≥98.0%
Leverantör: TCI EUROPE
Varning
F0158-25G F0158-250G
TCIAF0158-25GEA 350 SEK
TCIAF0158-25G TCIAF0158-250G
3-Fluorobenzoic acid ≥98.0%
3-Fluorobenzoic acid
Formel: C₇H₅FO₂
MW: 140,11 g/mol
Kokpunkt: 251 °C (1013 hPa)
Smältpunkt: 122 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002489
CAS nummer: 455-38-9
EINECS: 207-248-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4693aa4bdec1aa40f131ea540ccc5b3ac237bd69&vid=a635506563f0d3d434d97547e32d053f934c5810&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 121.0 to 125.0 deg-C
Solubility in Methanol very faint turbidity