3,4-Difluorobenzonitrile ≥98.0%

Leverantör: TCI EUROPE
Fara

D2341-25G D2341-5G
TCIAD2341-25GEA 1125 SEK
TCIAD2341-25G TCIAD2341-5G
3,4-Difluorobenzonitrile ≥98.0%
3,4-Difluorobenzonitrile
Formel: C₇H₃F₂N
MW: 139,1 g/mol
Kokpunkt: 95°C
Smältpunkt: 51 °C
MDL Number: MFCD00011666
CAS nummer: 64248-62-0
EINECS: 264-751-8
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7e9e78508b1cc4b1e2abf67cbc341117711c7e0&vid=cac9c5c582695c6316f521fe8729ff5a8cc0ab09&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 54.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR