2,5-Difluorobenzonitrile ≥98.0%
Leverantör: TCI EUROPE
Fara
D1834-25G D1834-5G
TCIAD1834-25GEA 3900 SEK
TCIAD1834-25G TCIAD1834-5G
2,5-Difluorobenzonitrile ≥98.0%
2,5-Difluorobenzonitrile
Formel: C₇H₃F₂N
MW: 139,1 g/mol
Kokpunkt: 92°C
Smältpunkt: 29,5…30.5 °C
MDL Number: MFCD00001777
CAS nummer: 64248-64-2
EINECS: 264-753-9
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbf21a84addf2034d8a08bf6aad8a6c32efd90ce&vid=bc33947be72d5f8f6a7009557c7bf059d4ddd208&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White or Colorles to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 98.0 %