2,4-Difluorobenzonitrile ≥98.0%

Leverantör: TCI
Varning

D1826-25G D1826-5G
TCIAD1826-25GEA 815 SEK
TCIAD1826-25G TCIAD1826-5G
2,4-Difluorobenzonitrile ≥98.0%
2,4-Difluorobenzonitrile
Formel: C₇H₃F₂N
Kokpunkt: 83°C
Smältpunkt: 46…47 °C
MDL Number: MFCD00009826
CAS nummer: 3939-09-1
EINECS: 223-523-8
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d78371ac7b3817e2f69351d5eb11b887088d60fb&vid=60c29bc577c8b7a7701f1516479668a394f67a01&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 46.0 to 49.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR