3,5-Difluorobenzaldehyde ≥98.0%
Leverantör: TCI EUROPE
Varning
D2626-25G D2626-5G
TCIAD2626-25GEA 2100 SEK
TCIAD2626-25G TCIAD2626-5G
3,5-Difluorobenzaldehyde ≥98.0%
3,5-Difluorobenzaldehyde
Formel: C₇H₄F₂O
MW: 142,1 g/mol
Kokpunkt: 61 - 63 °C (43 -rr)
Flampunkt: 56°C
Storage Temperature: Omgivande
MDL Number: MFCD00010329
CAS nummer: 32085-88-4
UN: 1325
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98b1ca28dca986d77bcd7dd823e76b754ac687de&vid=145e4f3395cab58a376dd4c7a579fddc027e5949&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %