2,6-Difluorobenzaldehyde ≥98.0%
Leverantör: TCI
|
Varning
|
D2452-25G
D2452-5G
TCIAD2452-25GEA
1700
SEK
TCIAD2452-25G
TCIAD2452-5G
2,6-Difluorobenzaldehyde ≥98.0%
2,6-Difluorobenzaldehyde
Formel:
C₇H₄F₂O MW: 142.1 g/mol Kokpunkt: 85°C Flampunkt: 73°C Storage Temperature: Omgivande |
MDL Number:
MFCD00010293 CAS nummer: 437-81-0 |
|
Specifikation testresultat
Appearance | White or Colorless to Yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |