3-Chlorothiophenol ≥97.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 3-Chlorobenzene-1-thiol, 1-Chloro-3-mercaptobenzene, 3-Chlorobenzenethiol, m-Chlorothiophenol

C1582-25G C1582-5G
TCIAC1582-25GEA 1100 SEK
TCIAC1582-25G TCIAC1582-5G
3-Chlorothiophenol ≥97.0%
3-Chlorothiophenol
Formel: C₆H₅ClS
MW: 144,62 g/mol
Kokpunkt: 88°C
Densitet: 1,2637 g/cm³ (12.5 °C)
Flampunkt: 90°C
MDL Number: MFCD00004839
CAS nummer: 2037-31-2
EINECS: 218-010-0
UN: 1760
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e482f4e807956ebcedede43e09776c1984359e8f&vid=0597b2c9536f9a28306b69d2ef4e5c306126a69f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR