2-Chlorothiophenol ≥97.0%

Leverantör: TCI EUROPE
Fara

Synonyms: o-Chlorothiophenol, 2-chlorothiophenol, 1-Chloro-2-mercaptobenzene, 2-Chlorobenzenethiol, 2-Chlorobenzene-1-thiol

C1975-25G C1975-5G
TCIAC1975-25GEA 1175 SEK
TCIAC1975-25G TCIAC1975-5G
2-Chlorothiophenol ≥97.0%
2-Chlorothiophenol
Formel: C₆H₅ClS
MW: 144,62 g/mol
Kokpunkt: 206°C
Flampunkt: 88°C
MDL Number: MFCD00004830
CAS nummer: 6320-03-2
EINECS: 228-667-5
UN: 1760
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=498e9179c1735c560f3380884b49dead66d77628&vid=498e9179c1735c560f3380884b49dead66d77628&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Iodometric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR