2,4,5-Triklorfenol ≥95.0%

Leverantör: TCI EUROPE
Varning

T0389-25G T0389-500G
TCIAT0389-25GEA 575 SEK
TCIAT0389-25G TCIAT0389-500G
2,4,5-Triklorfenol ≥95.0%
2,4,5-Triklorfenol
Formel: C₆H₃Cl₃O
MW: 197,45 g/mol
Kokpunkt: 115°C
Smältpunkt: 68 °C
Densitet: 1,68 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002170
CAS nummer: 95-95-4
EINECS: 202-467-8
UN: 2020
ADR: 6.1,III
Merck Index: 14,09643
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5fbbbffa95e5e7fab15cde0c920da3b8ffbefe5&vid=87f1d8cee9e1cf179b5992753feb0dd3f6b0ffbc&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 64.0 to 68.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR