2,3,6-Triklorfenol ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

T1114-1G T1114-10G
TCIAT1114-1GEA 615 SEK
TCIAT1114-1G TCIAT1114-10G
2,3,6-Triklorfenol ≥98.0% (by GC, titration analysis)
2,3,6-Triklorfenol
Formel: C₆H₃Cl₃O
MW: 197,45 g/mol
Kokpunkt: 250°C
Smältpunkt: 58 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002165
CAS nummer: 933-75-5
EINECS: 213-271-7
UN: 2020
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7a748e5b4d8379b3e137bc0d82f11c952e1a506&vid=b77ec4b4d7bfd14ec9d6cadbffc4c3576627aacb&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Gray to Red powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 54.0 to 57.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR