2,6-Diklorfenol ≥99.0%
Leverantör: TCI EUROPE
Fara
D0394-25G D0394-500G
TCIAD0394-25GEA 765 SEK
TCIAD0394-25G TCIAD0394-500G
2,6-Diklorfenol ≥99.0%
2,6-Diklorfenol
Formel: C₆H₄Cl₂O
MW: 163 g/mol
Kokpunkt: 80°C
Smältpunkt: 68 °C
Densitet: 1,653 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002176
CAS nummer: 87-65-0
EINECS: 201-761-3
UN: 2020
ADR: 6.1,III
Merck Index: 14,03073
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b19d92a090ed30412606957d88589da6ad7cfeb&vid=3bf6ff6aec19da63c4cb3d82cf2bba79f0b95f06&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Melting point 65.0 to 68.0 deg-C
Solubility in Methanol within almost transparency