4-Chloroanisole ≥98.0%
Leverantör: TCI
Synonyms:
1-Chloro-4-methoxybenzene
C0122-25G
C0122-500G
TCIAC0122-25GEA
295
SEK
TCIAC0122-25G
TCIAC0122-500G
4-Chloroanisole ≥98.0%
4-Chloroanisole
Formel:
C₇H₇ClO Kokpunkt: 198°C Smältpunkt: -18°C Densitet: 1,17 g/cm³ (20 °C) Flampunkt: 78°C Storage Temperature: Omgivande |
MDL Number:
MFCD00000616 CAS nummer: 623-12-1 EINECS: 210-772-2 |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...