4-Chloro-3-nitrophenol ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
C1529-25G C1529-5G
TCIAC1529-25GEA 845 SEK
TCIAC1529-25G TCIAC1529-5G
4-Chloro-3-nitrophenol ≥98.0% (by GC, titration analysis)
4-Chloro-3-nitrophenol
Formel: C₆H₄ClNO₃
MW: 173,56 g/mol
Smältpunkt: 125…128 °C
Densitet: 1 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00043546
CAS nummer: 610-78-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d5066a68525ff447985cd0fc401702438916e56&vid=6c3769f3db2121fc5149dd6df20134ea26c951e1&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 125.0 to 128.0 deg-C