2-Chloro-4-nitrophenol ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 1-Chloro-2-hydroxy-5-nitrobenzene

C0227-25G C0227-500G
TCIAC0227-25GEA 495 SEK
TCIAC0227-25G TCIAC0227-500G
2-Chloro-4-nitrophenol ≥98.0% (by GC, titration analysis)
2-Chloro-4-nitrophenol
Formel: C₆H₄ClNO₃
MW: 173,56 g/mol
Smältpunkt: 109…112 °C
MDL Number: MFCD00043910
CAS nummer: 619-08-9
EINECS: 210-578-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be08eeae40fd75ea58d40e3765639a0fe9cc1ec0&vid=8adf3f79bf259cbdd39ff8030cfc7bbb4af06e65&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 109.0 to 113.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR