1,3,5-Trichloro-2-nitrobenzene ≥96.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
2,4,6-Trichloronitrobenzene, 1-Nitro-2,4,6-trichlorobenzene
T0388-25G
T0388-5G
TCIAT0388-25GEA
855
SEK
TCIAT0388-25G
TCIAT0388-5G
1,3,5-Trichloro-2-nitrobenzene ≥96.0%
1,3,5-Trichloro-2-nitrobenzene
Formel:
C₆H₂Cl₃NO₂ MW: 226,45 g/mol Smältpunkt: 71 °C |
MDL Number:
MFCD00014690 CAS nummer: 18708-70-8 EINECS: 242-518-1 ADR: 6.1,III |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 70.0 to 74.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...