1-Klor-2-nitrobenzen ≥99.0%
Leverantör: TCI
Fara
C0220-25G C0220-500G
TCIAC0220-25GEA 215 SEK
TCIAC0220-25G TCIAC0220-500G
1-Klor-2-nitrobenzen ≥99.0%
1-Klor-2-nitrobenzen
Formel: C₆H₄ClNO₂
Kokpunkt: 246°C
Smältpunkt: 33 °C
Densitet: 1,368 g/cm³ (20 °C)
Flampunkt: 128 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007061
CAS nummer: 88-73-3
EINECS: 201-854-9
UN: 1578
ADR: 6.1,II
Merck Index: 14,02151
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54f0649753977dccba0df82d38f5cd4892c7213e&vid=087d922f26ed4054ea85dd72c46474582bf14b90&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White or Colorless to Yellow powder to lump to clear liquid
Purity (GC) min. 99.0 %
Solubility in Methanol almost transparency