1,4-Dichloro-2-nitrobenzene ≥99.0%
Leverantör: TCI EUROPE
Fara
D0387-25G D0387-500G
TCIAD0387-25GEA 215 SEK
TCIAD0387-25G TCIAD0387-500G
1,4-Dichloro-2-nitrobenzene ≥99.0%
1,4-Dichloro-2-nitrobenzene
Formel: C₆H₃Cl₂NO₂
MW: 192 g/mol
Kokpunkt: 98°C
Smältpunkt: 52…54 °C
Densitet: 1,704 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007074
CAS nummer: 89-61-2
EINECS: 201-923-3
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9fc3e5ece9e4ad2a72974aafe2b9a828e38e5451&vid=630edb9f746de45cec4981739a8cf3d35d62550a&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Yellow to Green powder to crystal
Purity (GC) min. 99.0 %
Melting point 54.0 to 57.0 deg-C
Solubility in Methanol almost transparency