1,3-Dichloro-4-nitrobenzene ≥99.0%
Leverantör: TCI EUROPE
Fara
Synonyms: 2,4-Dichloronitrobenzene
D0982-25G D0982-500G
TCIAD0982-25GEA 215 SEK
TCIAD0982-25G TCIAD0982-500G
1,3-Dichloro-4-nitrobenzene ≥99.0%
1,3-Dichloro-4-nitrobenzene
Formel: C₆H₃Cl₂NO₂
MW: 192 g/mol
Kokpunkt: 259°C
Smältpunkt: 28…31 °C
Densitet: 1,51 g/cm³ (20 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00007071
CAS nummer: 611-06-3
EINECS: 210-248-3
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=762d939ea882c1b449e45147207f61665dda0291&vid=865266beb6fa9a9c9a29b068b8e2a3b84759d18a&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White or Colorless to Orange to Green powder to lump to clear liquid
Purity (GC) min. 99.0 %
Solubility in Methanol almost transparency