2,5-Dikloranilin ≥98.0%
Leverantör: TCI EUROPE
Fara
D0322-25G D0322-500G D0322-100G
TCIAD0322-25GEA 295 SEK
TCIAD0322-25G TCIAD0322-500G TCIAD0322-100G
2,5-Dikloranilin ≥98.0%
2,5-Dikloranilin
Formel: C₆H₅Cl₂N
MW: 162,02 g/mol
Kokpunkt: 251 °C (1013 hPa)
Smältpunkt: 46…50 °C
Densitet: 1,54 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007667
CAS nummer: 95-82-9
EINECS: 202-455-2
UN: 3442
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e304dad89afee382916c9c9bb63a52f204ca9d5a&vid=ee64fb91b491c584880f71c2d58e30e72a186994&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 49.0 to 52.0 deg-C