2,3-Dikloranilin ≥98.0%
Leverantör: TCI EUROPE
Fara
D0320-25G D0320-500G
TCIAD0320-25GEA 285 SEK
TCIAD0320-25G TCIAD0320-500G
2,3-Dikloranilin ≥98.0%
2,3-Dikloranilin
Formel: C₆H₅Cl₂N
MW: 162,02 g/mol
Kokpunkt: 252 °C (1013 hPa)
Smältpunkt: 23…24 °C
Densitet: 1,383 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007657
CAS nummer: 608-27-5
EINECS: 210-157-9
UN: 3442
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7492094b80c88263372ebf2c5245d34af3c2e698&vid=2eff34ccd5ffb5810cfd7d80c9679d9e117392b9&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %