2,3,4-Trichloroaniline ≥98.0%

Leverantör: TCI EUROPE
Varning

T0798-25G T0798-5G
TCIAT0798-25GEA 6700 SEK
TCIAT0798-25G TCIAT0798-5G
2,3,4-Trichloroaniline ≥98.0%
2,3,4-Trichloroaniline
Formel: C₆H₄Cl₃N
MW: 196,46 g/mol
Kokpunkt: 292°C
Smältpunkt: 73 °C
Densitet: 1,54 g/cm³ (20 °C)
MDL Number: MFCD00042819
CAS nummer: 634-67-3
EINECS: 211-215-6
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0164939971fb5f725e71226a908cfdbd477fdc5&vid=6554dae5bf4d9d7f7c58cb3c02384d88b8aa8905&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 66.0 to 69.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR