1-Chloro-4-iodobenzene ≥99.0%

Leverantör: TCI EUROPE
Varning

C1075-25G C1075-250G
TCIAC1075-25GEA 430 SEK
TCIAC1075-25G TCIAC1075-250G
1-Chloro-4-iodobenzene ≥99.0%
1-Chloro-4-iodobenzene
Formel: C₆H₄ClI
MW: 238,45 g/mol
Kokpunkt: 227°C
Smältpunkt: 57 °C
Densitet: 1,886 g/cm³ (57 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00001053
CAS nummer: 637-87-6
EINECS: 211-305-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb6ea96ca0c38b31a6694987568988dd178abcdb&vid=dacd91d36703947daa3b2d7ae80316eb79569fed&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 99.0 %
Melting point 54.0 to 57.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR