1,3-Dichloro-5-iodobenzene ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 3,5-Dichloroiodobenzene

D1866-25G D1866-5G
TCIAD1866-25GEA 1525 SEK
TCIAD1866-25G TCIAD1866-5G
1,3-Dichloro-5-iodobenzene ≥98.0%
1,3-Dichloro-5-iodobenzene
Formel: C₆H₃Cl₂I
MW: 272,9 g/mol
Kokpunkt: 127 °C (20 -rr)
Smältpunkt: 57 °C
MDL Number: MFCD00001047
CAS nummer: 3032-81-3
EINECS: 221-216-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1b8306e33a4b9bd1497c02ead1db679dd72aab6&vid=28d1fd9656292e94168e7f255ee083e1456f2d35&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 55.0 to 59.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR