2-Chloro-4-fluorobenzaldehyde ≥97.0%
Leverantör: TCI
|
Varning
|
C1465-1G
C1465-5G
TCIAC1465-1GEA
230
SEK
TCIAC1465-1G
TCIAC1465-5G
2-Chloro-4-fluorobenzaldehyde ≥97.0%
2-Chloro-4-fluorobenzaldehyde
Formel:
C₇H₄ClFO Kokpunkt: 205 °C (1013 hPa) Smältpunkt: 60 °C |
MDL Number:
MFCD00042527 CAS nummer: 84194-36-5 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 60.0 to 63.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...