1,4-Diklor-2-fluorbenzen ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2,5-Dichlorofluorobenzene

D2513-25G D2513-5G
TCIAD2513-25GEA 1850 SEK
TCIAD2513-25G TCIAD2513-5G
1,4-Diklor-2-fluorbenzen ≥98.0%
1,4-Diklor-2-fluorbenzen
Formel: C₆H₃Cl₂F
MW: 164,99 g/mol
Kokpunkt: 168°C
Smältpunkt: 4 °C
Densitet: 1,383 g/cm³ (20 °C)
Flampunkt: 65°C
MDL Number: MFCD00060656
CAS nummer: 348-59-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3a658bfa54706f55d9d56f6fa5ec7ea48f1107c&vid=08d6f3b85ec3a3982698fc57a28fae14636f4c9c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR