4-Chloro-2-nitrobenzoic acid ≥98.0%
Leverantör: TCI
|
Varning
|
C0879-25G
C0879-500G
TCIAC0879-25GEA
1475
SEK
TCIAC0879-25G
TCIAC0879-500G
4-Chloro-2-nitrobenzoic acid ≥98.0%
4-Chloro-2-nitrobenzoic acid
Formel:
C₇H₄ClNO₄ Smältpunkt: 139…142 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00007214 CAS nummer: 6280-88-2 EINECS: 228-483-5 |
|
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 143.0 to 146.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...