2-Chloro-5-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Varning
C0877-25G C0877-500G
TCIAC0877-25GEA 215 SEK
TCIAC0877-25G TCIAC0877-500G
2-Chloro-5-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
2-Chloro-5-nitrobenzoic acid
Formel: C₇H₄ClNO₄
Smältpunkt: 164…167 °C
Densitet: 0,4 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007294
CAS nummer: 2516-96-3
EINECS: 219-739-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b993c4b13f6b68a61133eb6fe6f150c8dd6375a8&vid=554e37283eafe338b4228b0fdd504f443b83a4d8&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 164.0 to 168.0 deg-C