2-Amino-5-chlorobenzonitrile ≥98.0%

Leverantör: TCI
Fara

A1572-25G A1572-5G
TCIAA1572-25GEA 1875 SEK
TCIAA1572-25G TCIAA1572-5G
2-Amino-5-chlorobenzonitrile ≥98.0%
2-Amino-5-chlorobenzonitrile
Formel: C₇H₅ClN₂
Kokpunkt: 159°C
Smältpunkt: 96…98 °C
MDL Number: MFCD00017106
CAS nummer: 5922-60-1
EINECS: 227-651-5
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27582a1a962b8c2a73949353758d5152381bed94&vid=e108bb249c0171cd08c051e191c366d92bde7267&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 95.0 to 98.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR