2,5-Diklorbenzonitril ≥98.0%

Leverantör: TCI EUROPE
Fara

D4018-25G D4018-5G
TCIAD4018-25GEA 1275 SEK
TCIAD4018-25G TCIAD4018-5G
2,5-Diklorbenzonitril ≥98.0%
2,5-Diklorbenzonitril
Formel: C₇H₃Cl₂N
MW: 172,01 g/mol
Kokpunkt: 269 °C (1013 hPa)
Smältpunkt: 130…132 °C
MDL Number: MFCD00019741
CAS nummer: 21663-61-6
EINECS: 244-518-7
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1739cca8dba693bf9df08ccfe4c1319d2ac31a0&vid=d2067a9d385594ea6c0919243ad0bdd53ec5cccd&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 129.0 to 133.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR