2-Chloro-5-nitrobenzaldehyde ≥96.0%

Leverantör: TCI
Varning

C0219-25G C0219-250G
TCIAC0219-25GEA 415 SEK
TCIAC0219-25G TCIAC0219-250G
2-Chloro-5-nitrobenzaldehyde ≥96.0%
2-Chloro-5-nitrobenzaldehyde
Formel: C₇H₄ClNO₃
Smältpunkt: 75…78 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007293
CAS nummer: 6361-21-3
EINECS: 228-830-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b97823bddd00085217ae140ef83655b0cb900fdc&vid=b1f7650fe4dc2d48cc1b7bd357fd5d8a0e8aa8c7&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (Oxime Formation) min. 96.0 %
Melting point 78.0 to 81.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR