5-Chlorosalicylaldehyde ≥98.0% (by GC, titration analysis)

Leverantör: TCI
Varning

Synonyms: 5-Chloro-2-hydroxybenzaldehyde

C0738-25G C0738-5G
TCIAC0738-25GEA 1075 SEK
TCIAC0738-25G TCIAC0738-5G
5-Chlorosalicylaldehyde ≥98.0% (by GC, titration analysis)
5-Chlorosalicylaldehyde
Formel: C₇H₅ClO₂
Kokpunkt: 105 °C (12 -rr)
MDL Number: MFCD00003331
CAS nummer: 635-93-8
EINECS: 211-244-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ea2244af79557388c0177d6aa81cd65d3080a82&vid=0ea2244af79557388c0177d6aa81cd65d3080a82&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 99.0 to 104.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR