1,2,3-Triklorobenzen ≥98.0%

Leverantör: TCI EUROPE
Varning

T0377-25G T0377-500G
TCIAT0377-25GEA 215 SEK
TCIAT0377-25G TCIAT0377-500G
1,2,3-Triklorobenzen ≥98.0%
1,2,3-Triklorobenzen
Formel: C₆H₃Cl₃
MW: 181,45 g/mol
Kokpunkt: 219°C
Smältpunkt: 53…54 °C
Densitet: 1,45 g/cm³ (20 °C)
Flampunkt: 112 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000537
CAS nummer: 87-61-6
EINECS: 201-757-1
UN: 3077
ADR: 9,III
Merck Index: 14,09630
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=649e7d6977226245fb5f5ea6eb52b4bd2e572ed3&vid=e1accfaeb2bb550d3e25ac1abbf7eeab041bed53&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 52.0 to 56.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR