1,2,3,5-Tetrachlorobenzene ≥98.0%

Leverantör: TCI EUROPE
Varning

T0870-25G T0870-5G
TCIAT0870-25GEA 3375 SEK
TCIAT0870-25G TCIAT0870-5G
1,2,3,5-Tetrachlorobenzene ≥98.0%
1,2,3,5-Tetrachlorobenzene
Formel: C₆H₂Cl₄
MW: 215,89 g/mol
Kokpunkt: 246°C
Smältpunkt: 54 °C
Densitet: 1,858 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00000543
CAS nummer: 634-90-2
EINECS: 211-217-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11697feac037454913cd58c0d200d99eba7c6e31&vid=46c3ea501fe37a6f837db6724d063c62cf37a99f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 53.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR