Thiosalicylic acid ≥90.0%
Leverantör: TCI
|
Varning
|
Synonyms:
2-Thiobenzoic acid, 2-Mercaptobenzoic acid
M0054-25G
M0054-500G
TCIAM0054-25GEA
215
SEK
TCIAM0054-25G
TCIAM0054-500G
Thiosalicylic acid ≥90.0%
Thiosalicylic acid
Formel:
C₇H₆O₂S Kokpunkt: 230 °C (0,02 -rr) Smältpunkt: 165…166 °C Densitet: 1,489 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00004836 CAS nummer: 147-93-3 EINECS: 205-704-3 ADR: 9, Merck Index: 14,09360 |
|
Specifikation testresultat
Appearance | White to Light yellow to Green powder to crystal |
Purity (Argentmetric Titration) | min. 90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...