2,6-Dihydroxybenzoic acid ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
R0011-25G R0011-500G
TCIAR0011-25GEA 845 SEK
TCIAR0011-25G TCIAR0011-500G
2,6-Dihydroxybenzoic acid ≥98.0% (by GC, titration analysis)
2,6-Dihydroxybenzoic acid
Formel: C₇H₆O₄
MW: 154,12 g/mol
Kokpunkt: 339 °C (1013 hPa)
Smältpunkt: 152…158 °C
Densitet: 1,58 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002462
CAS nummer: 303-07-1
EINECS: 206-134-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2679196f72eaf03712f1678cd1aec64ce01c5846&vid=b73163bb3a95f5a5d648d882259b353567bd4b31&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Water very faint turbidity