5-Methoxysalicylic acid ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: 2-Hydroxy-5-methoxybenzoic acid
M1254-25G M1254-5G
TCIAM1254-25GEA 1250 SEK
TCIAM1254-25G TCIAM1254-5G
5-Methoxysalicylic acid ≥98.0% (by HPLC, titration analysis)
5-Methoxysalicylic acid
Formel: C₈H₈O₄
MW: 168,15 g/mol
Kokpunkt: 327 °C (1013 hPa)
Smältpunkt: 141…144 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002459
CAS nummer: 2612-02-4
EINECS: 220-037-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0bdb2622130f8fa88d5b66e003adc56b72bf8cc&vid=b0bdb2622130f8fa88d5b66e003adc56b72bf8cc&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 143.0 to 146.0 deg-C
Solubility in Methanol almost transparency