m-Anisic acid ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 3-Anisic acid, 3-Methoxybenzoic acid

A1070-25G A1070-250G
TCIAA1070-25GEA 365 SEK
TCIAA1070-25G TCIAA1070-250G
m-Anisic acid ≥98.0% (by GC, titration analysis)
m-Anisic acid
Formel: C₈H₈O₃
MW: 152,15 g/mol
Kokpunkt: 172°C
Smältpunkt: 103…106 °C
Densitet: 1,14 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002499
CAS nummer: 586-38-9
EINECS: 209-574-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=414ee0bbdde364e28494d44deed7d0d969d004e9&vid=c90c6d01fb6a01d85cb162e25276ffbfcca4be2d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 105.0 to 109.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR