o-Anisic acid ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 2-Methoxybenzoic acid

A0481-25G A0481-500G
TCIAA0481-25GEA 215 SEK
TCIAA0481-25G TCIAA0481-500G
o-Anisic acid ≥98.0% (by GC, titration analysis)
o-Anisic acid
Formel: C₈H₈O₃
MW: 152,15 g/mol
Kokpunkt: 280 °C (1013 hPa)
Smältpunkt: 99 °C
Densitet: 1,38 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002431
CAS nummer: 579-75-9
EINECS: 209-447-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=014aab838aa1202ae800d224c62c57d751c1dde9&vid=c3412493b429b93e26ec94c5e4e07aa8e4a03668&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 101.0 to 104.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR