2-Nitro-m-anisic acid ≥99.0%
Leverantör: TCI
Varning
M1036-1G M1036-10G
TCIAM1036-1GEA 365 SEK
TCIAM1036-1G TCIAM1036-10G
2-Nitro-m-anisic acid ≥99.0%
2-Nitro-m-anisic acid
Formel: C₈H₇NO₅
Kokpunkt: 388 °C (1013 hPa)
Smältpunkt: 174 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007143
CAS nummer: 4920-80-3
EINECS: 225-549-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a6662b2adc0a3404e418b3cad367e4db94e449a&vid=dde31a450de30b4b7ade70b77653ea5c08fe01db&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystaline
Purity (Neutralization titration) min. 99.0 %
Solubility in hot Methanol almost transparency