3,5-diaminobensoesyra ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
Varning
D0294-25G D0294-500G
TCIAD0294-25GEA 470 SEK
TCIAD0294-25G TCIAD0294-500G
3,5-diaminobensoesyra ≥98.0% (by HPLC, titration analysis)
3,5-diaminobensoesyra
Formel: C₇H₈N₂O₂
MW: 152,15 g/mol
Kokpunkt: 546 °C (1013 hPa)
Smältpunkt: 228°C
Densitet: 1,45 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007807
CAS nummer: 535-87-5
EINECS: 208-621-0
Merck Index: 14,02978
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=105764260764e4ba79e21262635ac8ab1a186796&vid=5769ee9cfa4c193abe39c48dbddeb5c7fa0ec11c&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Water very faint turbidity