3-Amino-4-hydroxybenzoic acid ≥97.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
Varning
A0859-25G A0859-5G
TCIAA0859-25GEA 5050 SEK
TCIAA0859-25G TCIAA0859-5G
3-Amino-4-hydroxybenzoic acid ≥97.0% (by HPLC, titration analysis)
3-Amino-4-hydroxybenzoic acid
Formel: C₇H₇NO₃
MW: 153,14 g/mol
Kokpunkt: 347 °C (1013 hPa)
Smältpunkt: 201 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007697
CAS nummer: 1571-72-8
EINECS: 216-390-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3aa36d801d6234b0cba36cecb91531e1ce9ae22&vid=dbc44df8171ab09f351025ef2a684f10b395c8ff&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Gray to Brown powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Solubility in hot Water within almost transparency