5-Amino-2-nitrobenzoic acid ≥98.0%

Leverantör: TCI
Varning

Synonyms: 2-Nitro-5-aminobenzoic acid, NSC 74455, 5-Amino-2-nitrobenzoic Acid, 3-Carboxy-4-nitroaniline

A0897-25G A0897-5G
TCIAA0897-25GEA 2925 SEK
TCIAA0897-25G TCIAA0897-5G
5-Amino-2-nitrobenzoic acid ≥98.0%
5-Amino-2-nitrobenzoic acid
Formel: C₇H₆N₂O₄
Kokpunkt: 481 °C (1013 hPa)
Smältpunkt: 234 °C
Storage Temperature: Omgivande
MDL Number: MFCD00035748
CAS nummer: 13280-60-9
EINECS: 236-283-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ae8922e6ffe8f779ff0f24c79e6beb29e76b8d6&vid=ce842c711cab7bd07fc42c2694e0178831c97b8c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR