5-Nitroanthranilic acid ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
2-Amino-5-nitrobenzoic acid
N0644-25G
N0644-5G
TCIAN0644-25GEA
935
SEK
TCIAN0644-25G
TCIAN0644-5G
5-Nitroanthranilic acid ≥98.0%
5-Nitroanthranilic acid
Formel:
C₇H₆N₂O₄ MW: 182,14 g/mol Smältpunkt: 278 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00017039 CAS nummer: 616-79-5 EINECS: 210-493-6 |
Specifikation testresultat
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...