4-Nitroanthranilic acid ≥97.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 2-Amino-4-nitrobenzoic acid

N0522-25G N0522-5G
TCIAN0522-25GEA 1150 SEK
TCIAN0522-25G TCIAN0522-5G
4-Nitroanthranilic acid ≥97.0% (by HPLC, titration analysis)
4-Nitroanthranilic acid
Formel: C₇H₆N₂O₄
MW: 182,14 g/mol
Smältpunkt: 271 °C
MDL Number: MFCD00007262
CAS nummer: 619-17-0
EINECS: 210-583-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0526b2ee6e9cfde78beee533d6af4076a9c794be&vid=29b84e74979642b68ada95a828d89278a22c4a1a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR