4-Cyanobenzoic acid ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

C0445-25G C0445-5G
TCIAC0445-25GEA 695 SEK
TCIAC0445-25G TCIAC0445-5G
4-Cyanobenzoic acid ≥98.0% (by GC, titration analysis)
4-Cyanobenzoic acid
Formel: C₈H₅NO₂
MW: 147,13 g/mol
Kokpunkt: 313 °C (1013 hPa)
Smältpunkt: 220…222 °C
Densitet: 1,382 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002528
CAS nummer: 619-65-8
EINECS: 210-606-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d150b7de58823b334fc48360916be15429c5989&vid=7f8a3eed3adf8c5562f28f790017b8164cbd2042&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR