Bensoesyra ≥99.0%
Leverantör: TCI
Fara
B2635-25G B2635-500G
TCIAB2635-25GEA 215 SEK
TCIAB2635-25G TCIAB2635-500G
Bensoesyra ≥99.0%
Bensoesyra
Formel: C₇H₆O₂
MW: 122.12 g/mol
Kokpunkt: 249°C
Flampunkt: 121 (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00002398
CAS nummer: 65-85-0
EINECS: 200-618-2
REACH: 01-2119455536-33
Merck Index: 14,01091
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b69e5adfb69ad7f73c3020d9529a6a2d96caea29&vid=e68397769f25b4dd4553d3f45ab5069369c40f2e&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 122.0 to 126.0 deg-C
Solubility in NH3ap. (2+3) almost transparency