Phenoxyacetone ≥97.0%

Leverantör: TCI
Varning

Synonyms: 1-Phenoxypropan-2-one

P0112-25G P0112-5G
TCIAP0112-25GEA 3675 SEK
TCIAP0112-25G TCIAP0112-5G
Phenoxyacetone ≥97.0%
Phenoxyacetone
Formel: (C₆H₅)OCH₂C(O)CH₃
Kokpunkt: 230°C
Densitet: 1,0852 g/cm³ (20 °C)
Flampunkt: 85°C
MDL Number: MFCD00008767
CAS nummer: 621-87-4
EINECS: 210-712-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c25323102cf70ac168ee1f4a571f899f16519c88&vid=e154b3813c340760c370cb52fde7b854c1622dd3&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR