2,5-Dimethoxybenzonitrile ≥98.0%

Leverantör: TCI EUROPE
Fara

D3659-1G D3659-5G
TCIAD3659-1GEA 215 SEK
TCIAD3659-1G TCIAD3659-5G
2,5-Dimethoxybenzonitrile ≥98.0%
2,5-Dimethoxybenzonitrile
Formel: C₉H₉NO₂
MW: 163,18 g/mol
Kokpunkt: 283 °C (1013 hPa)
Smältpunkt: 82 °C
MDL Number: MFCD00016375
CAS nummer: 5312-97-0
EINECS: 226-169-2
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c51876edf5a733bdd2cc4d2cd68e12257124a899&vid=623e814f2bbb678cc3a44d80e361d099277c1d52&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 81.0 to 85.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR